Assay Details
| Gene Symbol: AT5G24750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122384.3 | NP_568452.2 | 9 - 10 | 881 | 73 | |
| GenBank mRNA | KJ676981.1 | - | 9 - 10 | 738 | 73 | |
More Information
Set Membership: |
|

AT5G24750
Genomic Map