Assay Details
| Gene Symbol: RRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122415.2 | NP_568464.1 | 15 - 16 | 2627 | 88 | |
| GenBank mRNA | AF360212.1 | - | 15 - 16 | 2627 | 88 | |
| AY040060.1 | - | 15 - 16 | 2394 | 88 | ||
More Information
Set Membership: |
|

RRC1
Genomic Map