Assay Details
| Gene Symbol: RRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122415.2 | NP_568464.1 | 16 - 17 | 2783 | 84 | |
| GenBank mRNA | AF360212.1 | - | 16 - 17 | 2783 | 84 | |
| AY040060.1 | - | 16 - 17 | 2550 | 84 | ||
More Information
Set Membership: |
|

RRC1
Genomic Map