Assay Details
| Gene Symbol: AT5G25080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122417.3 | NP_197890.1 | 2 - 3 | 402 | 119 | |
| GenBank mRNA | AF378870.1 | - | 2 - 3 | 367 | 119 | |
| AY050469.1 | - | 2 - 3 | 282 | 119 | ||
| AY087408.1 | - | 2 - 3 | 399 | 119 | ||
More Information
Set Membership: |
|

AT5G25080
Genomic Map