Assay Details
| Gene Symbol: AT5G25080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122417.3 | NP_197890.1 | 4 - 5 | 538 | 92 | |
| GenBank mRNA | AF378870.1 | - | 4 - 5 | 503 | 92 | |
| AY050469.1 | - | 4 - 5 | 418 | 92 | ||
More Information
Set Membership: |
|

AT5G25080
Genomic Map