Assay Details
| Gene Symbol: CID6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343938.1 | NP_001331123.1 | 1 - 2 | 697 | 106 | |
| NM_122465.4 | NP_197936.1 | 2 - 3 | 561 | 106 | ||
| GenBank mRNA | AK316708.1 | - | 2 - 3 | 485 | 106 | |
| BT010477.1 | - | 1 - 2 | 370 | 106 | ||
More Information
Set Membership: |
|

CID6
Genomic Map