Assay Details
| Gene Symbol: PEX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036862.2 | NP_001031939.1 | 2 - 3 | 550 | 104 | |
| NM_122477.2 | NP_568476.1 | 3 - 4 | 357 | 104 | ||
| GenBank mRNA | AY084297.1 | - | 3 - 4 | 357 | 104 | |
| BT025616.1 | - | 1 - 2 | 262 | 104 | ||
| DQ027035.1 | - | 1 - 2 | 262 | 104 | ||
More Information
Set Membership: |
|

PEX4
Genomic Map