Assay Details
| Gene Symbol: EIF3B-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122479.4 | NP_568477.1 | 1 - 2 | 267 | 143 | |
| GenBank mRNA | BT002393.1 | - | 1 - 2 | 219 | 143 | |
More Information
Set Membership: |
|

EIF3B-2
Genomic Map