Assay Details
| Gene Symbol: NADP-ME3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343951.1 | NP_001331266.1 | 9 - 10 | 1101 | 63 | |
| NM_001343952.1 | NP_001331265.1 | 9 - 10 | 1101 | 63 | ||
| NM_122489.3 | NP_197960.1 | 9 - 10 | 1125 | 63 | ||
More Information
Set Membership: |
|

NADP-ME3
Genomic Map