Assay Details
| Gene Symbol: FC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036864.1 | NP_001031941.1 | 2 - 3 | 565 | 137 | |
| NM_122504.5 | NP_197975.3 | 2 - 3 | 574 | 137 | ||
| GenBank mRNA | AK226466.1 | - | 2 - 3 | 407 | 137 | |
| X73417.1 | - | 416 | 137 | |||
More Information
Set Membership: |
|

FC1
Genomic Map