Assay Details
| Gene Symbol: CLC-D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343969.1 | NP_001330038.1 | 5 - 6 | 882 | 82 | |
| NM_122525.3 | NP_197996.1 | 5 - 6 | 897 | 82 | ||
More Information
Set Membership: |
|

CLC-D
Genomic Map