Assay Details
Gene Symbol: AT5G26260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122527.4 | NP_568483.1 | 4 - 5 | 671 | 106 | |
GenBank mRNA | AF370133.1 | - | 4 - 5 | 463 | 106 | |
AY051023.1 | - | 4 - 5 | 450 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
