Assay Details
| Gene Symbol: CBP60G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343987.1 | NP_001331616.1 | 4 - 5 | 1032 | 110 | |
| NM_122574.4 | NP_198044.2 | 4 - 5 | 1035 | 110 | ||
More Information
Set Membership: |
|

CBP60G
Genomic Map