Assay Details
| Gene Symbol: CBP60G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203479.1 | NP_001190408.1 | 5 - 6 | 1353 | 66 | |
| NM_122574.4 | NP_198044.2 | 5 - 6 | 1380 | 66 | ||
More Information
Set Membership: |
|

CBP60G
Genomic Map