Assay Details
| Gene Symbol: AGL93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122577.2 | NP_198047.1 | 1 | 754 | 118 | |
| GenBank mRNA | AY141228.1 | - | 1 | 732 | 118 | |
| BT028900.1 | - | 1 | 732 | 118 | ||
More Information
Set Membership: |
|

AGL93
Genomic Map