Assay Details
| Gene Symbol: SYP41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036873.2 | NP_001031950.1 | 2 - 3 | 968 | 85 | |
| NM_001343989.1 | NP_001331924.1 | 2 - 3 | 707 | 85 | ||
| NM_122580.4 | NP_198050.1 | 2 - 3 | 968 | 85 | ||
| GenBank mRNA | AF067789.2 | - | 2 - 3 | 561 | 85 | |
More Information
Set Membership: |
|

SYP41
Genomic Map