Assay Details
| Gene Symbol: SYP41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036873.2 | NP_001031950.1 | 4 - 5 | 1122 | 155 | |
| NM_001343989.1 | NP_001331924.1 | 4 - 5 | 861 | 155 | ||
| NM_122580.4 | NP_198050.1 | 4 - 5 | 1122 | 155 | ||
| GenBank mRNA | AF067789.2 | - | 4 - 5 | 715 | 155 | |
More Information
Set Membership: |
|

SYP41
Genomic Map