Assay Details
| Gene Symbol: TPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203480.1 | NP_001190409.1 | 13 - 14 | 1875 | 80 | |
| NM_122585.5 | NP_198055.3 | 14 - 15 | 1955 | 80 | ||
| GenBank mRNA | BT004037.1 | - | 14 - 15 | 1786 | 80 | |
| BT005165.1 | - | 14 - 15 | 1653 | 80 | ||
More Information
Set Membership: |
|

TPR3
Genomic Map