Assay Details
| Gene Symbol: emb2204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102059.2 | NP_564157.1 | 4 - 5 | 528 | 80 | |
| GenBank mRNA | AX765880.1 | - | 4 - 5 | 515 | 80 | |
More Information
Set Membership: |
|

emb2204
Genomic Map