Assay Details
| Gene Symbol: AGL104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102063.3 | NP_173632.1 | 2 - 3 | 323 | 121 | |
| GenBank mRNA | DQ056457.1 | - | 2 - 3 | 237 | 121 | |
More Information
Set Membership: |
|

AGL104
Genomic Map