Assay Details
| Gene Symbol: AUF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102072.4 | NP_173641.1 | 1 | 1120 | 77 | |
| GenBank mRNA | AK176237.1 | - | 1 | 977 | 77 | |
| BT012651.1 | - | 1 | 885 | 77 | ||
More Information
Set Membership: |
|

AUF2
Genomic Map