Assay Details
| Gene Symbol: UGT85A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102085.3 | NP_173652.1 | 1 - 2 | 544 | 110 | |
| GenBank mRNA | DQ446278.1 | - | 1 - 2 | 516 | 110 | |
| KJ138994.1 | - | 1 - 2 | 516 | 110 | ||
More Information
Set Membership: |
|

UGT85A7
Genomic Map