Assay Details
| Gene Symbol: ATL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102099.2 | NP_173666.1 | 1 | 1233 | 66 | |
| GenBank mRNA | AK226235.1 | - | 1 | 1139 | 66 | |
| BT011235.1 | - | 1 | 1210 | 66 | ||
More Information
Set Membership: |
|

ATL15
Genomic Map