Assay Details
| Gene Symbol: RABG3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102121.3 | NP_173688.1 | 6 - 7 | 716 | 60 | |
| GenBank mRNA | AB071850.1 | - | 6 - 7 | 559 | 60 | |
| BT024589.1 | - | 6 - 7 | 559 | 60 | ||
| Y09821.1 | - | 706 | 60 | |||
More Information
Set Membership: |
|

RABG3B
Genomic Map