Assay Details
| Gene Symbol: PAB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102123.4 | NP_173690.1 | 5 - 6 | 1831 | 77 | |
| GenBank mRNA | AY054490.1 | - | 5 - 6 | 1830 | 77 | |
More Information
Set Membership: |
|

PAB3
Genomic Map