Assay Details
| Gene Symbol: TH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102141.3 | NP_173707.2 | 3 - 4 | 501 | 94 | |
| GenBank mRNA | BT020417.1 | - | 3 - 4 | 456 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TH1| Gene Symbol: TH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102141.3 | NP_173707.2 | 3 - 4 | 501 | 94 | |
| GenBank mRNA | BT020417.1 | - | 3 - 4 | 456 | 94 | |
Set Membership: |
|