Assay Details
| Gene Symbol: TH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102141.3 | NP_173707.2 | 7 - 8 | 1092 | 103 | |
| GenBank mRNA | BT020417.1 | - | 7 - 8 | 1047 | 103 | |
More Information
Set Membership: |
|

TH1
Genomic Map