Assay Details
| Gene Symbol: AT1G23030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102151.5 | NP_173716.1 | 1 - 2 | 519 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G23030| Gene Symbol: AT1G23030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102151.5 | NP_173716.1 | 1 - 2 | 519 | 113 | |
Set Membership: |
|