Assay Details
| Gene Symbol: AST91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332586.1 | NP_001320230.1 | 8 - 9 | 2219 | 103 | |
| NM_001332587.1 | NP_001320231.1 | 9 - 10 | 2134 | 103 | ||
| NM_001332588.1 | NP_001320232.1 | 9 - 10 | 2089 | 103 | ||
| NM_102157.4 | NP_173722.1 | 10 - 11 | 2018 | 103 | ||
More Information
Set Membership: |
|

AST91
Genomic Map