Assay Details
| Gene Symbol: AST91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332588.1 | NP_001320232.1 | 1 - 2 | 514 | 69 | |
| NM_102157.4 | NP_173722.1 | 1 - 2 | 528 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001332588.1 transcripts.
Set Membership: |
|

AST91
Genomic Map