Assay Details
| Gene Symbol: AST91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332586.1 | NP_001320230.1 | 1 - 2 | 983 | 81 | |
| NM_001332587.1 | NP_001320231.1 | 2 - 3 | 898 | 81 | ||
| NM_001332588.1 | NP_001320232.1 | 2 - 3 | 853 | 81 | ||
| NM_102157.4 | NP_173722.1 | 3 - 4 | 782 | 81 | ||
More Information
Set Membership: |
|

AST91
Genomic Map