Assay Details
| Gene Symbol: AST91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332586.1 | NP_001320230.1 | 2 - 3 | 1159 | 74 | |
| NM_001332587.1 | NP_001320231.1 | 3 - 4 | 1074 | 74 | ||
| NM_001332588.1 | NP_001320232.1 | 3 - 4 | 1029 | 74 | ||
| NM_102157.4 | NP_173722.1 | 4 - 5 | 958 | 74 | ||
More Information
Set Membership: |
|

AST91
Genomic Map