Assay Details
| Gene Symbol: AST91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332586.1 | NP_001320230.1 | 3 - 4 | 1220 | 66 | |
| NM_001332587.1 | NP_001320231.1 | 4 - 5 | 1135 | 66 | ||
| NM_001332588.1 | NP_001320232.1 | 4 - 5 | 1090 | 66 | ||
| NM_102157.4 | NP_173722.1 | 5 - 6 | 1019 | 66 | ||
More Information
Set Membership: |
|

AST91
Genomic Map