Assay Details
| Gene Symbol: AST91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332586.1 | NP_001320230.1 | 5 - 6 | 1455 | 69 | |
| NM_001332587.1 | NP_001320231.1 | 6 - 7 | 1370 | 69 | ||
| NM_001332588.1 | NP_001320232.1 | 6 - 7 | 1325 | 69 | ||
| NM_102157.4 | NP_173722.1 | 7 - 8 | 1254 | 69 | ||
More Information
Set Membership: |
|

AST91
Genomic Map