Assay Details
| Gene Symbol: AT1G23280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102177.3 | NP_173742.1 | 7 - 8 | 877 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G23280| Gene Symbol: AT1G23280 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102177.3 | NP_173742.1 | 7 - 8 | 877 | 83 | |
Set Membership: |
|