Assay Details
| Gene Symbol: CAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102189.4 | NP_173754.2 | 1 - 2 | 1809 | 142 | |
| GenBank mRNA | AY062732.1 | - | 1 - 2 | 677 | 142 | |
| AY114649.1 | - | 1 - 2 | 673 | 142 | ||
More Information
Set Membership: |
|

CAF2
Genomic Map