Assay Details
| Gene Symbol: CAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102189.4 | NP_173754.2 | 2 - 3 | 1935 | 115 | |
| GenBank mRNA | AY062732.1 | - | 2 - 3 | 803 | 115 | |
| AY114649.1 | - | 2 - 3 | 799 | 115 | ||
More Information
Set Membership: |
|

CAF2
Genomic Map