Assay Details
| Gene Symbol: CAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102189.4 | NP_173754.2 | 5 - 6 | 2438 | 65 | |
| GenBank mRNA | AY062732.1 | - | 5 - 6 | 1306 | 65 | |
| AY114649.1 | - | 5 - 6 | 1302 | 65 | ||
More Information
Set Membership: |
|

CAF2
Genomic Map