Assay Details
| Gene Symbol: AT1G23410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102190.3 | NP_173755.1 | 1 | 485 | 96 | |
| GenBank mRNA | AY085389.1 | - | 1 | 414 | 96 | |
| BT024763.1 | - | 1 | 398 | 96 | ||
| Gene Symbol: CAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102189.4 | NP_173754.2 | 1 | 540 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_102189.4 transcripts.
Set Membership: |
|

AT1G23410
Genomic Map