Assay Details
| Gene Symbol: AT1G24200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102266.1 | NP_173830.1 | 2 - 3 | 323 | 133 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G24200| Gene Symbol: AT1G24200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102266.1 | NP_173830.1 | 2 - 3 | 323 | 133 | |
Set Membership: |
|