Assay Details
| Gene Symbol: AT1G24230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102269.1 | NP_173833.1 | 2 - 3 | 351 | 86 | |
| GenBank mRNA | BT031359.1 | - | 2 - 3 | 351 | 86 | |
More Information
Set Membership: |
|

AT1G24230
Genomic Map