Assay Details
| Gene Symbol: TNO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198156.2 | NP_001185085.1 | 1 - 2 | 304 | 91 | |
| NM_102291.5 | NP_173855.5 | 1 - 2 | 106 | 91 | ||
More Information
Set Membership: |
|

TNO1
Genomic Map