Assay Details
| Gene Symbol: TNO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198156.2 | NP_001185085.1 | 6 - 7 | 5065 | 79 | |
| NM_102291.5 | NP_173855.5 | 4 - 5 | 5092 | 79 | ||
More Information
Set Membership: |
|

TNO1
Genomic Map