Assay Details
| Gene Symbol: THO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198157.1 | NP_001185086.1 | 11 - 12 | 1017 | 127 | |
| NM_001332655.1 | NP_001321786.1 | 10 - 11 | 1208 | 127 | ||
| NM_001332656.1 | NP_001321785.1 | 10 - 11 | 1208 | 127 | ||
| NM_001332657.1 | NP_001321784.1 | 11 - 12 | 1121 | 127 | ||
| NM_001332658.1 | NP_001321783.1 | 11 - 12 | 958 | 127 | ||
| NM_001332659.1 | NP_001321782.1 | 10 - 11 | 1168 | 127 | ||
| NM_001332660.1 | NP_001321781.1 | 9 - 10 | 1243 | 127 | ||
| NM_001332661.1 | NP_001321780.1 | 5 - 6 | 540 | 127 | ||
| NM_102309.6 | NP_173871.6 | 11 - 12 | 1017 | 127 | ||
More Information
Set Membership: |
|

THO2
Genomic Map