Assay Details
| Gene Symbol: THO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198157.1 | NP_001185086.1 | 14 - 15 | 1363 | 75 | |
| NM_001332655.1 | NP_001321786.1 | 13 - 14 | 1554 | 75 | ||
| NM_001332656.1 | NP_001321785.1 | 13 - 14 | 1554 | 75 | ||
| NM_001332657.1 | NP_001321784.1 | 14 - 15 | 1467 | 75 | ||
| NM_001332658.1 | NP_001321783.1 | 14 - 15 | 1304 | 75 | ||
| NM_001332659.1 | NP_001321782.1 | 13 - 14 | 1514 | 75 | ||
| NM_001332660.1 | NP_001321781.1 | 12 - 13 | 1589 | 75 | ||
| NM_001332661.1 | NP_001321780.1 | 8 - 9 | 886 | 75 | ||
| NM_102309.6 | NP_173871.6 | 14 - 15 | 1363 | 75 | ||
More Information
Set Membership: |
|

THO2
Genomic Map