Assay Details
| Gene Symbol: THO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198157.1 | NP_001185086.1 | 20 - 21 | 2186 | 104 | |
| NM_001332655.1 | NP_001321786.1 | 19 - 20 | 2377 | 104 | ||
| NM_001332656.1 | NP_001321785.1 | 19 - 20 | 2377 | 104 | ||
| NM_001332657.1 | NP_001321784.1 | 20 - 21 | 2290 | 104 | ||
| NM_001332658.1 | NP_001321783.1 | 20 - 21 | 2127 | 104 | ||
| NM_001332659.1 | NP_001321782.1 | 19 - 20 | 2337 | 104 | ||
| NM_001332660.1 | NP_001321781.1 | 18 - 19 | 2412 | 104 | ||
| NM_001332661.1 | NP_001321780.1 | 14 - 15 | 1709 | 104 | ||
| NM_102309.6 | NP_173871.6 | 20 - 21 | 2186 | 104 | ||
More Information
Set Membership: |
|

THO2
Genomic Map