Assay Details
| Gene Symbol: THO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198157.1 | NP_001185086.1 | 29 - 30 | 3819 | 85 | |
| NM_001332655.1 | NP_001321786.1 | 28 - 29 | 4010 | 85 | ||
| NM_001332656.1 | NP_001321785.1 | 28 - 29 | 4010 | 85 | ||
| NM_001332657.1 | NP_001321784.1 | 29 - 30 | 3923 | 85 | ||
| NM_001332658.1 | NP_001321783.1 | 29 - 30 | 3760 | 85 | ||
| NM_001332659.1 | NP_001321782.1 | 28 - 29 | 3970 | 85 | ||
| NM_001332660.1 | NP_001321781.1 | 27 - 28 | 4045 | 85 | ||
| NM_001332661.1 | NP_001321780.1 | 23 - 24 | 3342 | 85 | ||
| NM_102309.6 | NP_173871.6 | 29 - 30 | 3819 | 85 | ||
More Information
Set Membership: |
|

THO2
Genomic Map