Assay Details
| Gene Symbol: THO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198157.1 | NP_001185086.1 | 31 - 32 | 4411 | 85 | |
| NM_001332655.1 | NP_001321786.1 | 30 - 31 | 4602 | 85 | ||
| NM_001332656.1 | NP_001321785.1 | 30 - 31 | 4602 | 85 | ||
| NM_001332657.1 | NP_001321784.1 | 31 - 32 | 4515 | 85 | ||
| NM_001332658.1 | NP_001321783.1 | 31 - 32 | 4352 | 85 | ||
| NM_001332660.1 | NP_001321781.1 | 29 - 30 | 4637 | 85 | ||
| NM_001332661.1 | NP_001321780.1 | 25 - 26 | 3934 | 85 | ||
| NM_102309.6 | NP_173871.6 | 31 - 32 | 4411 | 85 | ||
More Information
Set Membership: |
|

THO2
Genomic Map