Assay Details
| Gene Symbol: THO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198157.1 | NP_001185086.1 | 8 - 9 | 758 | 71 | |
| NM_001332655.1 | NP_001321786.1 | 7 - 8 | 949 | 71 | ||
| NM_001332656.1 | NP_001321785.1 | 7 - 8 | 949 | 71 | ||
| NM_001332657.1 | NP_001321784.1 | 8 - 9 | 862 | 71 | ||
| NM_001332658.1 | NP_001321783.1 | 8 - 9 | 699 | 71 | ||
| NM_001332659.1 | NP_001321782.1 | 7 - 8 | 909 | 71 | ||
| NM_001332660.1 | NP_001321781.1 | 6 - 7 | 984 | 71 | ||
| NM_001332661.1 | NP_001321780.1 | 2 - 3 | 281 | 71 | ||
| NM_102309.6 | NP_173871.6 | 8 - 9 | 758 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001332661.1 transcripts.
Set Membership: |
|

THO2
Genomic Map