Assay Details
| Gene Symbol: AT1G25375 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102348.4 | NP_564232.1 | 5 - 6 | 1028 | 84 | |
| GenBank mRNA | AK175321.1 | - | 5 - 6 | 847 | 84 | |
More Information
Set Membership: |
|

AT1G25375
Genomic Map